Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ ImmunoCAP™ Venom Allergens

Catalog No. 14452501P
Change view
Click to view available options
Quantity:
10 Tests
16 Tests
Code:
Ri205
i1
i2
i3
i4
i5
i75
Common Name:
Bumblebee
Common wasp (Yellow jacket)
European hornet
Honey bee
Paper wasp
White-faced hornet
Yellow hornet
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Code Common Name
14-452-501P 16 Tests i3 Common wasp (Yellow jacket)
14-414-301P 16 Tests i1 Honey bee
14-422-301P 16 Tests i5 Yellow hornet
14-422-101P 16 Tests i2 White-faced hornet
14-434-001P 10 Tests i75 European hornet
14-452-601P 16 Tests i4 Paper wasp
14-527-010P 10 Tests Ri205 Bumblebee
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 14-452-501P Supplier Thermo Scientific™ Supplier No. 14452501
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Measure specifc IgE antibodies using ImmunoCAP™ allergens, an essential tool to aid in the diagnosis of allergic conditions.

Quantify specific IgE antibodies with the ImmunoCAP™ Venom Allergens, essential tools to aid in the diagnosis of allergic conditions. IgE antibodies appear in human serum and plasma as a result of sensitization to a specific allergen. Measurement of circulating IgE antibodies provides an objective assessment of sensitization to an allergen. In general, low IgE antibody levels indicate a low probability of allergic disease, whereas high antibody levels to an allergen show a higher correlation with an allergic disease.

ImmunoCAP Specific IgE offers the laboratory:
  • Truly quantitative measuring range from 0.1 to 100 kUA/L
  • Intra-assay CV (%) comparable to routine clinical chemistry immunoassays
  • Excellent consistency over time, and between countries, systems, labs and persons
Expected test values
ImmunoCAP Specific IgE detects IgE antibodies in the range 0.1 to 100 kUA /l, where A represents allergen-specific antibodies. The result is reported quantitatively. In clinical practice, 0.35 kUA /l has commonly been used as a cut-off. A large number of studies have been performed to evaluate the clinical performance of ImmunoCAP Specific IgE tests in allergy diagnosis. Clinical performance is expressed as sensitivity, ranging from 84 to 95%, and specificity, ranging from 85 to 94%. Sensitivity and specificity were reported from multi-center studies including several hundred patients tested for a range of different allergens.
  • Specimen collection and preparation
  • Use serum and plasma (EDTA or heparin) samples from venous or capillary blood
  • Collect blood samples using standard procedures
  • Keep specimens at room temperature (RT) for shipping purposes only
  • Store at 2–8°C up to one week, otherwise store at –20°C
  • Avoid repeated freezing and thawing

Note: Blood samples for testing ImmunoCAP drugs and venom should be collected during or close to the event, preferably not later than 6 months after exposure. If the test result is negative and an IgE-mediated reaction is still strongly suspected, it is advisable to draw a new sample and repeat the test at 5 to 6 weeks.

The presence of specific IgE antibodies is useful to identify the allergens that elicit symptoms and signs of allergy in patients with respiratory allergic diseases including asthma, food allergy, and anaphylactic sensitivity.

Knowing IgE antibody levels provides guidance to clinicians to:
  • Identify offending allergens
  • Tailor advice to individual patients
  • Indicate development of tolerance (food allergy, specific immunotherapy)
  • Inform optimized individual medical treatment plans
  • Refer to specialists appropriately
  • Evaluate whether or not specific immunotherapy is an option

Specific IgE testing also aids in the identification of patients at risk of:
  • The allergy march – a progression of skin symptoms to respiratory symptoms
  • Severe symptoms including asthma exacerbations – a progression of mild symptoms to severe symptoms
  • Progression of recurrent symptoms to persistent symptoms

As in all diagnostic testing, a definitive clinical diagnosis should be made by the clinician after evaluation of all clinical and laboratory findings.
TRUSTED_SUSTAINABILITY

Specifications

Scientific Name Vespula spp.
Regulatory Status FDA Cleared
Code Type Venom
Common Name Common wasp (Yellow jacket)
Quantity 16 Tests
Code i3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.