Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ ImmunoCAP™ Weed Pollen Allergens

Catalog No. 14502510P Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 Tests
16 Tests
Code:
Rw203
Rw204
Rw207
Rw23
Rw45
Rw46
Rw82
w1
w10
w11
w12
w13
w14
w15
w16
w17
w18
w19
w2
w20
w21
w3
w4
w5
w6
w7
w8
w9
Common Name:
Alfalfa
Careless weed
Cocklebur
Common pigweed
Common ragweed
Dandelion
Dog fennel
False ragweed
Firebush (Kochia)
Giant ragweed
Goldenrod
Goosefoot, Lamb’s quarters
Lupin
Marguerite, Ox-eye daisy
Mugwort
Nettle
Plantain (English), Ribwort
Rape
Rough marshelder
Saltwort (prickly), Russian thistle
Scale, Lenscale
Sheep sorrel
Sunflower
Wall pellitory
Western ragweed
Wormwood
Yellow dock
28 product options available for selection
Product selection table with 28 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Code Common Name
14-502-510P 10 Tests Rw204 Sunflower
14-410-401P 16 Tests w21 Wall pellitory
14-410-301P 16 Tests w6 Mugwort
14-415-501P 16 Tests w8 Dandelion
14-415-301P 16 Tests w3 Giant ragweed
14-415-401P 16 Tests w7 Marguerite, Ox-eye daisy
14-415-201P 16 Tests w2 Western ragweed
14-415-601P 16 Tests w9 Plantain (English), Ribwort
14-415-701P 16 Tests w10 Goosefoot, Lamb’s quarters
14-418-601P 16 Tests w19 Wall pellitory
14-424-701P 16 Tests w4 False ragweed
14-424-801P 16 Tests w5 Wormwood
14-425-201P 16 Tests w14 Common pigweed
14-424-901P 16 Tests w11 Saltwort (prickly), Russian thistle
14-425-001P 16 Tests w12 Goldenrod
14-425-301P 16 Tests w15 Scale, Lenscale
14-425-101P 16 Tests w13 Cocklebur
14-425-501P 16 Tests w17 Firebush (Kochia)
14-425-401P 16 Tests w16 Rough marshelder
14-425-601P 16 Tests w18 Sheep sorrel
14-425-701P 16 Tests w20 Nettle
14-495-010P 10 Tests Rw23 Yellow dock
14-495-110P 10 Tests Rw45 Alfalfa
14-495-310P 10 Tests Rw82 Careless weed
14-495-210P 10 Tests Rw46 Dog fennel
14-502-410P 10 Tests Rw203 Rape
14-502-210P 10 Tests Rw207 Lupin
14-416-201P 16 Tests w1 Common ragweed
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 28 options available.
28 options
This item is not returnable. View return policy
Catalog No. 14-502-510P Supplier Thermo Scientific™ Supplier No. 14502510
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Measure specifc IgE antibodies using ImmunoCAP™ allergens, an essential tool to aid in the diagnosis of allergic conditions.

Quantify specific IgE antibodies with ImmunoCAP™ Weed Pollen Allergens, essential tools to aid in the diagnosis of allergic conditions. IgE antibodies appear in human serum and plasma as a result of sensitization to a specific allergen. Measurement of circulating IgE antibodies provides an objective assessment of sensitization to an allergen. In general, low IgE antibody levels indicate a low probability of allergic disease, whereas high antibody levels to an allergen show a higher correlation with an allergic disease.

ImmunoCAP Specific IgE offers the laboratory:
  • Truly quantitative measuring range from 0.1 to 100 kUA/L
  • Intra-assay CV (%) comparable to routine clinical chemistry immunoassays
  • Excellent consistency over time, and between countries, systems, labs and persons
Expected test values
ImmunoCAP Specific IgE detects IgE antibodies in the range 0.1 to 100 kUA /l, where A represents allergen-specific antibodies. The result is reported quantitatively. In clinical practice, 0.35 kUA /l has commonly been used as a cut-off. A large number of studies have been performed to evaluate the clinical performance of ImmunoCAP Specific IgE tests in allergy diagnosis. Clinical performance is expressed as sensitivity, ranging from 84 to 95%, and specificity, ranging from 85 to 94%. Sensitivity and specificity were reported from multi-center studies including several hundred patients tested for a range of different allergens.
  • Specimen collection and preparation
  • Use serum and plasma (EDTA or heparin) samples from venous or capillary blood
  • Collect blood samples using standard procedures
  • Keep specimens at room temperature (RT) for shipping purposes only
  • Store at 2–8°C up to one week, otherwise store at –20°C
  • Avoid repeated freezing and thawing

Note: Blood samples for testing ImmunoCAP drugs and venom should be collected during or close to the event, preferably not later than 6 months after exposure. If the test result is negative and an IgE-mediated reaction is still strongly suspected, it is advisable to draw a new sample and repeat the test at 5 to 6 weeks.

The presence of specific IgE antibodies is useful to identify the allergens that elicit symptoms and signs of allergy in patients with respiratory allergic diseases including asthma, food allergy, and anaphylactic sensitivity.

Knowing IgE antibody levels provides guidance to clinicians to:
  • Identify offending allergens
  • Tailor advice to individual patients
  • Indicate development of tolerance (food allergy, specific immunotherapy)
  • Inform optimized individual medical treatment plans
  • Refer to specialists appropriately
  • Evaluate whether or not specific immunotherapy is an option

Specific IgE testing also aids in the identification of patients at risk of:
  • The allergy march – a progression of skin symptoms to respiratory symptoms
  • Severe symptoms including asthma exacerbations – a progression of mild symptoms to severe symptoms
  • Progression of recurrent symptoms to persistent symptoms

As in all diagnostic testing, a definitive clinical diagnosis should be made by the clinician after evaluation of all clinical and laboratory findings.
TRUSTED_SUSTAINABILITY

Specifications

Scientific Name Helianthus annuus
Regulatory Status ASR
Code Type Weed
Common Name Sunflower
Quantity 10 Tests
Code Rw204
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.