Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meridian Bioscience™ ImmunoCard STAT!™ FLU A&B
GSA_VA

Catalog No. 23315126 Shop All Meridian Bioscience Products
Change view
Click to view available options
Quantity:
32 Tests (30 Test + 2 Controls)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-315-126 32 Tests (30 Test + 2 Controls)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-315-126 Supplier Meridian Bioscience™ Supplier No. 782030

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

32 test devices, 32 Buffer ampules, 30 Flocked swabs, 1 Positive and 1 negative control swabs

Meridian Bioscience ImmunoCard™ STAT! FLU A&B is a rapid Flu test for Nasal and NP swabs

  • CLIA waived 
  • Results in 10 to 15 minutes
  • Meets new FDA regulations performance criteria.

Order Info

EA (kit of 32 tests)

Warranty and Services

FDA cleared

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Certifications/Compliance FDA cleared
Content And Storage 2°C to 30°C
Clia Complexity Waived
Detectable Analytes Influenza A and B
Format Immunocard
Sensitivity Flu A: 89.2% Flu B: 86.4%
For Use With (Application) Nasal and nasopharyngeal swabs
Packaging Type 1 Kit
Quantity 32 Tests (30 Test + 2 Controls)
Specificity Flu A: 99.4%; Flu B: 99.0%
Test Time 10 to 15 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.