Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meridian Bioscience™ ImmunoCard STAT!™ FLU A&B Accessory Kit
GSA_VA

Catalog No. 23315127 Shop All Meridian Bioscience Products
Change view
Click to view available options
Quantity:
30 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-315-127 30 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-315-127 Supplier Meridian Bioscience™ Supplier No. 781130
 Restricted Item
Add to Cart
Add to Cart

Includes

60 transfer pipets, 1 bottle of buffer

Requires

ImmunoCard STAT! FLU A&B kit

Meridian Bioscience ImmunoCard™ STAT! FLU A&B Accessory Kit for testing nasal aspirate/washes and samples in transport media

  • For testing nasal aspirate/washes and samples in transport media

Order Info

EA (kit of 30 tests)

Certifications

FDA cleared

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Certifications/Compliance FDA cleared
Content And Storage 2°C to 30°C
Clia Complexity Moderate
Format Immunocard
Sensitivity Flu A: 89.2% Flu B: 86.4%
For Use With (Application) ImmunoCard STAT! FLU A&B kit
Includes 60 transfer pipets, 1 Buffer bottle
Packaging Type EA (box)
Quantity 30 Tests
Specificity Flu A: 99.4%; Flu B: 99.0%
Test Time 10 to 15 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.