Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meridian Bioscience™ ImmunoCard STAT! HpSA
GSA_VA

Catalog No. 23315121 Shop All Meridian Bioscience Products
Change view
Click to view available options
Quantity:
20 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-315-121 20 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-315-121 Supplier Meridian Bioscience™ Supplier No. 750220
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Diagnose, monitor, and confirm eradication for all phases of H. pylori patient management. ImmunoCard STAT! HpSA is a clia-waived, easy-to-use, active infection test for the diagnosis of H. pylori.

  • A rapid in vitro qualitative procedure for the detection of Helicobacter pylori antigens in human stool
  • The stool antigen detection is intended to aid in the diagnosis of H. pylori infection and to demonstrate loss of H. pylori stool antigen following treatment

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Content And Storage 2°C to 8°C
CE Marker Yes
Detectable Analytes H. pylori Antigen
Format Immunocard
Type Rapid Immunochromatographic Lateral Flow
Includes 20 tests each, sample diluent, positive control, transfer pipettes
Quantity 20 Tests
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.