Learn More
Biocare Medical, LLC Immunocare TBS Wash Buffer, 10X
SDP
Catalog No. 5083295
Encompass_Preferred
Immunocare TBS Wash Buffer (sodium azide and thimerasol free) works well for manual and automated immunohistochemistry applications as well as other laboratory procedures requiring a high quality TBS buffer with superior pH stability. The buffer contains surfactant and allows for reagents to spread more uniformly across tissue sections on slides. It has been specifically formulated with Enzyme Activator for Alkaline Phosphatase detection systems. It can also be used on an automated staining system.
Catalog No. 50-832-95 Supplier Biocare Medical, LLC Supplier No. TWB943M
Immunocare TBS Wash Buffer (sodium azide and thimerasol free) works well for manual and automated immunohistochemistry applications as well as other laboratory procedures requiring a high quality TBS buffer with superior pH stability. The buffer contains surfactant and allows for reagents to spread more uniformly across tissue sections on slides. It has been specifically formulated with Enzyme Activator for Alkaline Phosphatase detection systems. It can also be used on an automated staining system.