Learn More
STEMCELL Technologies ImmunoCult™ Dendritic Cell Maturation Supplement, Size: 0.5 mL
SDP
Catalog No. NC1439543
Encompass
Use ImmunoCult™ Dendritic Cell Maturation Supplement for the maturation of immature dendritic cells (DCs) to mature DCs.For your convenience, the medium is available either individually or as part of the ImmunoCult™ Dendritic Cell Culture Kit (/products/immunocult-dendritic-cell-culture-kit.html). The ImmunoCult™ Dendritic Cell Maturation Supplement is formulated for use in combination with ImmunoCult™-ACF Dendritic Cell Medium (/products/immunocult-acf-dendritic-cell-medium.html) and ImmunoCult™-ACF Dendritic Cell Differentiation Supplement (/products/immunocult-acf-dendritic-cell-differentiation-supplement.html) to support the maturation of immature DCs.For more information on protocols for maturation of DCs using ImmunoCult™ Dendritic Cell Maturation Supplement, please explorethe Product Information Sheet (PIS) (/products/immunocult-acf-dendritic-cell-medium.html#section-protocols-and-documentation).
Catalog No. NC1439543 Supplier STEMCELL Technologies Supplier No. 10989
Use ImmunoCult™ Dendritic Cell Maturation Supplement for the maturation of immature dendritic cells (DCs) to mature DCs.For your convenience, the medium is available either individually or as part of the ImmunoCult™ Dendritic Cell Culture Kit (/products/immunocult-dendritic-cell-culture-kit.html). The ImmunoCult™ Dendritic Cell Maturation Supplement is formulated for use in combination with ImmunoCult™-ACF Dendritic Cell Medium (/products/immunocult-acf-dendritic-cell-medium.html) and ImmunoCult™-ACF Dendritic Cell Differentiation Supplement (/products/immunocult-acf-dendritic-cell-differentiation-supplement.html) to support the maturation of immature DCs.For more information on protocols for maturation of DCs using ImmunoCult™ Dendritic Cell Maturation Supplement, please explorethe Product Information Sheet (PIS) (/products/immunocult-acf-dendritic-cell-medium.html#section-protocols-and-documentation).