Learn More
STEMCELL Technologies ImmunoCult™ Human CD3/CD28/CD2 T Cell Activator
SDP
Catalog No. NC2748367
Encompass
Achieve activation and expansion of T cells without magnetic beads, feeder cells, or antigens. This product’s gentle activation stimulus ensures a high viability of activated T cells, which can be expanded in ImmunoCult™-XF T Cell Expansion Medium (Catalog #10981) or other media for human T cells. ImmunoCult™ Human CD3/CD28/CD2 T Cell Activator consists of soluble antibody complexes that bind to and cross-link CD3, CD28, and CD2 cell surface ligands, thereby providing the required primary and co-stimulatory signals for T cell activation.
Catalog No. NC2748367 Supplier STEMCELL Technologies Supplier No. 1000785
Achieve activation and expansion of T cells without magnetic beads, feeder cells, or antigens. This product’s gentle activation stimulus ensures a high viability of activated T cells, which can be expanded in ImmunoCult™-XF T Cell Expansion Medium (Catalog #10981) or other media for human T cells. ImmunoCult™ Human CD3/CD28/CD2 T Cell Activator consists of soluble antibody complexes that bind to and cross-link CD3, CD28, and CD2 cell surface ligands, thereby providing the required primary and co-stimulatory signals for T cell activation.