Learn More
STEMCELL Technologies ImmunoCult™ Human Th1 Differentiation Supplement, Size: 1 mL
SDP
Catalog No. NC1381584
Encompass
ImmunoCult™ Human Th1 Differentiation Supplement contains a combination of human cytokines and a blocking antibody formulated to promote the robust activation, expansion, and differentiation of peripheral blood-derived, naïve, CD4+ human T cells into Th1 cells. ImmunoCult™ Human Th1 Differentiation Supplement is intended for use with ImmunoCult™-XF T Cell Expansion Medium (Catalog #10981) and ImmunoCult™ Human CD3/CD28 T Cell Activator (Catalog #10971). This product is recommended for:• Research into the regulation of human naïve T cell differentiation and Th1 function• Development of procedures to expand and polarize naïve T cells toward a Th1 subset
This item is not returnable. View return policy
Catalog No. NC1381584 Supplier STEMCELL Technologies Supplier No. 10973
ImmunoCult™ Human Th1 Differentiation Supplement contains a combination of human cytokines and a blocking antibody formulated to promote the robust activation, expansion, and differentiation of peripheral blood-derived, naïve, CD4+ human T cells into Th1 cells. ImmunoCult™ Human Th1 Differentiation Supplement is intended for use with ImmunoCult™-XF T Cell Expansion Medium (Catalog #10981) and ImmunoCult™ Human CD3/CD28 T Cell Activator (Catalog #10971). This product is recommended for:• Research into the regulation of human naïve T cell differentiation and Th1 function• Development of procedures to expand and polarize naïve T cells toward a Th1 subset