Learn More
STEMCELL Technologies ImmunoCult™ Human Th2 Differentiation Supplement, Size: 1 mL
SDP
Catalog No. NC1381586
Encompass
ImmunoCult™ Human Th2 Differentiation Supplement contains a human cytokine and blocking antibody formulated to promote the robust activation, expansion, and differentiation of peripheral blood-derived, naïve, CD4+ human T cells into Th2 cells. ImmunoCult™ Human Th2 Differentiation Supplement is intended for use with ImmunoCult™-XF T Cell Expansion Medium and ImmunoCult™ Human CD3/CD28 T Cell Activator. This product is recommended for:• Research into the regulation of human naïve T cell differentiation and Th2 function• Development of procedures to expand and polarize naïve T cells toward a Th2 subset
Catalog No. NC1381586 Supplier STEMCELL Technologies Supplier No. 10975
ImmunoCult™ Human Th2 Differentiation Supplement contains a human cytokine and blocking antibody formulated to promote the robust activation, expansion, and differentiation of peripheral blood-derived, naïve, CD4+ human T cells into Th2 cells. ImmunoCult™ Human Th2 Differentiation Supplement is intended for use with ImmunoCult™-XF T Cell Expansion Medium and ImmunoCult™ Human CD3/CD28 T Cell Activator. This product is recommended for:• Research into the regulation of human naïve T cell differentiation and Th2 function• Development of procedures to expand and polarize naïve T cells toward a Th2 subset