Learn More
STEMCELL Technologies ImmunoCult™ Human Treg Differentiation Supplement, Size: 1 mL
SDP
Catalog No. NC1327611
Encompass
ImmunoCult™ Human Treg Differentiation Supplement contains a human cytokine and small molecule formulated to promote the robust activation, expansion, and differentiation of peripheral blood-derived, naïve, CD4+ human T cells into regulatory T cells (Tregs). ImmunoCult™ Human Treg Differentiation Supplement is intended for use with ImmunoCult™-XF T Cell Expansion Medium and ImmunoCult™ Human CD3/CD28 T Cell Activator. This product is recommended for:• Research into the regulation of human naïve T cell differentiation and Treg function• Development of procedures to expand and polarize naïve T cells into Tregs
This item is not returnable. View return policy
Catalog No. NC1327611 Supplier STEMCELL Technologies Supplier No. 10977
ImmunoCult™ Human Treg Differentiation Supplement contains a human cytokine and small molecule formulated to promote the robust activation, expansion, and differentiation of peripheral blood-derived, naïve, CD4+ human T cells into regulatory T cells (Tregs). ImmunoCult™ Human Treg Differentiation Supplement is intended for use with ImmunoCult™-XF T Cell Expansion Medium and ImmunoCult™ Human CD3/CD28 T Cell Activator. This product is recommended for:• Research into the regulation of human naïve T cell differentiation and Treg function• Development of procedures to expand and polarize naïve T cells into Tregs