Learn More
Description
Includes
- Cutters for 1 and 2mm troughs
- Dies for two well patterns
- Leveling table
- Immuno frame holder
- Immuno frames, six each
- Staining and rinsing tanks, eight each
- Suction needle
- 2.5 x 7.6cm glass slides, pack of 144
- Gel knife
- 2.5 x 6.4cm microporous wicks, pack of 600
- High resolution buffer, 12 x 18g
- Noble Agar, 114g
- Allen wrench
- Contains all the required materials to perform immunoelectrophoresis, except for the electrophoresis system and power supply
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.