Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Immunoelectrophoresis Kit
SDPSCI_ED

Catalog No. S90247ND Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S90247ND 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S90247ND Supplier Edvotek™ Supplier No. 272
Add to Cart
Add to Cart

Includes

  • IgG
  • Whole serum
  • Albumin
  • Antibody to whole serum
  • Antibody to IgG
  • UltraSpec-Agarose™
  • Electrophoresis buffer (25X concentrate)
  • 50 Microtest tubes
  • Two 10mL pipets
  • 20 Filter papers
  • 10 Well cutters
  • 10 60mm Petri plates

Requires

  • Horizontal electrophoresis apparatus
  • Power supply
  • Automatic micropipet and tips (5-50μL)
  • Waterbath (set to 55°C)
  • Microscope slides (1 x 3 in.)
  • Distilled water
  • 400 to 600mL Beaker
  • 1000mL Graduated cylinder
  • Bunsen burner, hotplate, or microwave
  • Container with lid (large enough to hold electrophoresis trays)
  • Paper toweling
  • Plastic wrap or foil
  • 37°C Incubation oven

Intermediate level experiment uses immunoelectrophoresis to separate and characterize a mixture of proteins.

  • Lets students examine the specificity of the antigen-antibody interaction
  • Identifies proteins based on their combined electrophoretic and immunological properties
  • Experiment uses agarose gel electrophoresis

No experiment components contain human serum.

Specifications

Grade 9 to Undergraduate
Includes Instructions, proteins, antibodies; reagents, agarose, buffer, transfer pipets, well cutters, tubes, filter paper, 10 mL pipettes, and 60 mm petri plates
Quantity 10 separations
Requires Horizontal electrophoresis apparatus, power supply, automatic micropipettes with tips, microscope slides (1 x 3 in.), distilled water, 400-600 mL beaker, 1000 mL graduated cylinder, microwave, humidifying chamber, paper towels, plastic wrap or foil, and 37°C incubation oven (optional)
Time Range Electrophoresis: 60 minutes; Incubation: overnight
Content And Storage Refrigerator
Class Size 10 Lab Groups
Product Type Electrophoresis Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.