Learn More
Electron Microscopy Sciences EMS Immunofluorescence 2-Chamber
SDP
Catalog No. 5025782
Encompass_Preferred
A two well incubation chamber for use in immunocytochemistry (by Miguel Berrios, SUNY Stony Brook). Also available in a one well version. This chamber accepts regular microscope slides or coverslips and is assembled without any bonding substrates. Antibody incubation and washing are done in the chamber wells. It is then taken apart before microscopic examination. It is a proven solution to contamination spills, non-specific fluorescent signals, and sample dehydration in indirect immunofluorescence assays. Drastically reduces non-specific backgrounds.
Overall measurement: 4½"(L) x 2"(W) x 7/16"(T) (113 x 51 x 12 mm). Cavity: ¾" (dia.) x ¼"(D) (20 mm x 6 mm).
Catalog No. 50-257-82 Supplier Electron Microscopy Sciences Supplier No. 70330
A two well incubation chamber for use in immunocytochemistry (by Miguel Berrios, SUNY Stony Brook). Also available in a one well version. This chamber accepts regular microscope slides or coverslips and is assembled without any bonding substrates. Antibody incubation and washing are done in the chamber wells. It is then taken apart before microscopic examination. It is a proven solution to contamination spills, non-specific fluorescent signals, and sample dehydration in indirect immunofluorescence assays. Drastically reduces non-specific backgrounds.
Overall measurement: 4½"(L) x 2"(W) x 7/16"(T) (113 x 51 x 12 mm). Cavity: ¾" (dia.) x ¼"(D) (20 mm x 6 mm).