Learn More
Biomatik Corporation Immunoglobulin E (Ige) Monoclonal Antibody
Supplier:  Biomatik Corporation CAU30524
Encompass_Preferred
Immunoglobulin E (Ige) Monoclonal Antibody is a Pig (Sus scrofa; Porcine) Monoclonal Antibody to Immunoglobulin E (IgE). The Immunoglobulin E (Ige) Monoclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Mouse. Species: Pig (Sus scrofa; Porcine). Immunogen: Asp393~Gly410. Applications: WB, IHC, ICC, IP. Target Synonyms: Immunoglobulin E (IgE). Accession Number: K7ZJQ0. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5220