Learn More
Sigma Aldrich Fine Chemicals Biosciences Immunoglobulin G huma100MG
Catalog No. 502900819
Encompass
Immunoglobulin G (IgG) subtype is the most abundant serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids. IgG provides protection from infections caused by bacteria fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defence of the neonate against infections Human IgG is purified from normal human serum and contains 1.0% sodium and 15% water.
Catalog No. 50-290-0819 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 56834100MG
Add to Cart
Add to Cart
Immunoglobulin G (IgG) subtype is the most abundant serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids. IgG provides protection from infections caused by bacteria fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defence of the neonate against infections Human IgG is purified from normal human serum and contains 1.0% sodium and 15% water.