Learn More
Cusabio Technology LLC Immunoglobulin G IgG
Catalog No. 5000052030
Encompass_Preferred
This kit detects IgG in sheep serum plasma cell culture supernates and tissue homogenates Designed for quantitative analysis it employs a competitive ELISA method with a broad detection range of 0 9375 g/ml to 240 g/ml and a sensitivity of 0 9375 g/ml ensuring precise measurement of low to high IgG concentrations The assay can be completed within 1-5 hours requiring only 50-100 l of sample per test Results are determined spectrophotometrically at 450 nm providing reliable and reproducible data for research or diagnostic applications Its compatibility with diverse biological samples makes it a versatile tool for studying immunoglobulin G levels in experimental or clinical settings
Catalog No. 50-000-52030 Supplier Cusabio Technology LLC Supplier No. CSBE14400SH
This kit detects IgG in sheep serum plasma cell culture supernates and tissue homogenates Designed for quantitative analysis it employs a competitive ELISA method with a broad detection range of 0 9375 g/ml to 240 g/ml and a sensitivity of 0 9375 g/ml ensuring precise measurement of low to high IgG concentrations The assay can be completed within 1-5 hours requiring only 50-100 l of sample per test Results are determined spectrophotometrically at 450 nm providing reliable and reproducible data for research or diagnostic applications Its compatibility with diverse biological samples makes it a versatile tool for studying immunoglobulin G levels in experimental or clinical settings