Learn More
Biomatik Corporation Immunoglobulin G (Igg) Monoclonal Antibody
Supplier:  Biomatik Corporation CAU29256
Encompass_Preferred
Immunoglobulin G (Igg) Monoclonal Antibody is a Rat (Rattus norvegicus) Monoclonal Antibody to Immunoglobulin G (IgG). The Immunoglobulin G (Igg) Monoclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Mouse. Species: Rat (Rattus norvegicus). Applications: WB, IHC, ICC, IP. Target Synonyms: Immunoglobulin G (IgG). Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5855