Learn More
Athens Research And Technology Inc Immunoglobulin G1 (IgG1), Native Human Plasma, ≥95% (SDS-PAGE), 1 mg
Catalog No. NC3884614
Encompass_Preferred
Immunoglobulin G1 (IgG1) is a native protein purified from normal human plasma with ≥95% (SDS-PAGE), supplied frozen in a 1 mg pack. Molecular weight: 146 kDa. Common research applications include immunology, antibody characterization and assay development.
Catalog No. NC3884614 Supplier Athens Research And Technology Inc Supplier No. 16160907071
Immunoglobulin G1 (IgG1) is a native protein purified from normal human plasma with ≥95% (SDS-PAGE), supplied frozen in a 1 mg pack. Molecular weight: 146 kDa. Common research applications include immunology, antibody characterization and assay development.