Learn More
Athens Research And Technology Inc Immunoglobulin G2, Native, ≥95% (SDS-PAGE), 1 mg
Catalog No. NC3884615
Encompass_Preferred
Immunoglobulin G2 is a native protein with ≥95% (SDS-PAGE), supplied frozen in a 1 mg pack. Molecular weight: 146 kDa. Common research applications include immunology, antibody characterization and assay development.
Catalog No. NC3884615 Supplier Athens Research And Technology Inc Supplier No. 16160907072
Immunoglobulin G2 is a native protein with ≥95% (SDS-PAGE), supplied frozen in a 1 mg pack. Molecular weight: 146 kDa. Common research applications include immunology, antibody characterization and assay development.