Learn More
Diagnostic Biosystems IMMUNOHISTO-SEALER ImmunoHisto-Sealer™
Catalog No. 502407357
Encompass_Preferred
ImmunoHisto-Sealer™ is a preservation solution that acts as a sealant to improve tissue morphology and stain over an extended period of time. When applied to the stained section, specimens can be permanently mounted without chromogens fading. Additional benefits of the ImmunoHisto-Sealer™ include: saves hands on time when mounting slides, avoids the use of toxic chemicals such as xylene and eliminates accumulation of hazardous waste.\n\nThis product is designed for tissue specimens stained with peroxidase, alkaline phosphatase-based systems and various chromogens. After treating with ImmunoHisto-Sealer, water solution chromogens can be mounted in Permanent Mounting Media.
Catalog No. 50-240-7357 Supplier Diagnostic Biosystems Supplier No. K0761000
ImmunoHisto-Sealer™ is a preservation solution that acts as a sealant to improve tissue morphology and stain over an extended period of time. When applied to the stained section, specimens can be permanently mounted without chromogens fading. Additional benefits of the ImmunoHisto-Sealer™ include: saves hands on time when mounting slides, avoids the use of toxic chemicals such as xylene and eliminates accumulation of hazardous waste.\n\nThis product is designed for tissue specimens stained with peroxidase, alkaline phosphatase-based systems and various chromogens. After treating with ImmunoHisto-Sealer, water solution chromogens can be mounted in Permanent Mounting Media.