Learn More
Electron Microscopy Sciences ImmunoHistoZyme™ Ready-to-Use 15 ML
SDP
Catalog No. 501926531
Encompass_Preferred
ImmunoHistoZyme is used for proteolytic digestion of formalin-fixed paraffin-embedded (FFPE) tissue sections prior to application of antibodies. In IHC most commonly used fixative like formalin mask tissue antigens (cellular, membrane and nuclear) by cross-linking process, this results in poor or no staining in IHC. ImmunoHistoZyme digestion of FFPE tissue section improves accessibility of antibodies to tissue antigens.
Store in temperature of 2-8 degree C. DO NOT FREEZE. Use as antigen retriever for some antibodies in IHC. Please refer to primary antibody protocol for IHC. Reagent: Ready-to-use ImmunoHistoZyme Reagent.
Catalog No. 50-192-6531 Supplier Electron Microscopy Sciences Supplier No. 6414213
ImmunoHistoZyme is used for proteolytic digestion of formalin-fixed paraffin-embedded (FFPE) tissue sections prior to application of antibodies. In IHC most commonly used fixative like formalin mask tissue antigens (cellular, membrane and nuclear) by cross-linking process, this results in poor or no staining in IHC. ImmunoHistoZyme digestion of FFPE tissue section improves accessibility of antibodies to tissue antigens.
Store in temperature of 2-8 degree C. DO NOT FREEZE. Use as antigen retriever for some antibodies in IHC. Please refer to primary antibody protocol for IHC. Reagent: Ready-to-use ImmunoHistoZyme Reagent.