Learn More
STEMCELL Technologies EasySep™ Human Plasmacytoid DC Isolation Kit, For processing: 2 x 10^9 cells
SDP
Catalog No. 502036746
Encompass
The EasySep™ Human Plasmacytoid DC Isolation Kit is designed to isolate plasmacytoid dendritic cells (pDCs) from human peripheral blood mononuclear cells (PBMCs) or leukapheresis by immunomagnetic negative selection. The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles. The magnetically labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube.This product replaces the EasySep™ Human Plasmacytoid DC Enrichment Kit (Catalog #19062), for even faster cell isolations and is compatible with a greater number of EasySep™ magnets.
Catalog No. 50-203-6746 Supplier STEMCELL Technologies Supplier No. 17977
The EasySep™ Human Plasmacytoid DC Isolation Kit is designed to isolate plasmacytoid dendritic cells (pDCs) from human peripheral blood mononuclear cells (PBMCs) or leukapheresis by immunomagnetic negative selection. The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles. The magnetically labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube.This product replaces the EasySep™ Human Plasmacytoid DC Enrichment Kit (Catalog #19062), for even faster cell isolations and is compatible with a greater number of EasySep™ magnets.