Learn More
STEMCELL Technologies EasySep™ Release Mouse APC Positive Selection Kit, For processing: 1 x 10^9 cells
SDP
Catalog No. 502045472
Encompass
The EasySep™ Release Mouse APC Positive Selection Kit is designed to isolate cells that are labeled with APC-conjugated antibodies from mouse tissue by immunomagnetic positive selection.Desired cells are labeled with antibodies and magnetic particles, and separated without columns using an EasySep™ magnet. Unwanted cells are simply poured off, while desired cells remain in the tube. Then, bound magnetic particles are removed from the EasySep™-isolated, APC-antibody labeled cells, which are immediately available for downstream applications. Following cell isolation with this EasySep™ Release kit, antibody complexes remain bound to the cell surface and may interact with Brilliant Violet™ antibody conjugates, polyethylene glycol-modified proteins or other chemically related ligands.
Catalog No. 50-204-5472 Supplier STEMCELL Technologies Supplier No. 1000033
The EasySep™ Release Mouse APC Positive Selection Kit is designed to isolate cells that are labeled with APC-conjugated antibodies from mouse tissue by immunomagnetic positive selection.Desired cells are labeled with antibodies and magnetic particles, and separated without columns using an EasySep™ magnet. Unwanted cells are simply poured off, while desired cells remain in the tube. Then, bound magnetic particles are removed from the EasySep™-isolated, APC-antibody labeled cells, which are immediately available for downstream applications. Following cell isolation with this EasySep™ Release kit, antibody complexes remain bound to the cell surface and may interact with Brilliant Violet™ antibody conjugates, polyethylene glycol-modified proteins or other chemically related ligands.