Learn More
Alkali Scientific ASI™ Immunoplate 8-Well Strip, Flat Bottom, PS, No Lid, 100/Cs, White, Maxibinding
SDP
Catalog No. 503027168
Encompass_Preferred
▫White plates: maximum reflection and minimum crosstalk for luminescence assay. ▫ Black plates: minimum light scattering and crosstalk during fluorescence assay.  ▫ Maxibinding : Modified polystyrene surface for higher binding capacity for proteins and other                           molecules with both hydrophilic and hydrophobic regions.                            (Recommended for most ELISA experiments) ▫ Unibinding : Suitable for adsorption of hydrophobic molecules. (Antigen ELISA) ▪ Suitable for fluorescence assay, luminescence assay▪ 2 type surface▪ 8well strip type▪ Lid not included▪ Flat bottom▪ Raised rims on the wells▪ Alphanumeric labeling▪ Designed for easy handling and automation
Catalog No. 50-302-7168 Supplier Alkali Scientific Supplier No. TSMW08
▫White plates: maximum reflection and minimum crosstalk for luminescence assay. ▫ Black plates: minimum light scattering and crosstalk during fluorescence assay.  ▫ Maxibinding : Modified polystyrene surface for higher binding capacity for proteins and other                           molecules with both hydrophilic and hydrophobic regions.                            (Recommended for most ELISA experiments) ▫ Unibinding : Suitable for adsorption of hydrophobic molecules. (Antigen ELISA) ▪ Suitable for fluorescence assay, luminescence assay▪ 2 type surface▪ 8well strip type▪ Lid not included▪ Flat bottom▪ Raised rims on the wells▪ Alphanumeric labeling▪ Designed for easy handling and automation