Learn More
Alkali Scientific Immunoplate, 96-Well, PS, Black, MaxiBinding, 10/pk, 100/cs
SDP
Catalog No. 502389516
Encompass_Preferred
Suitable for fluorescence and luminescence assay. 8 well strip type. No lid. Raised rims on the wells. Alphanumeric labeling. 85.4 x 127.6 x 14.4 mm, Bottom Surface Area: 0.33cm2, Total Volume: 400µl, Working Volume: 350µl.
Catalog No. 50-238-9516 Supplier Alkali Scientific Supplier No. TMB1096
Suitable for fluorescence and luminescence assay. 8 well strip type. No lid. Raised rims on the wells. Alphanumeric labeling. 85.4 x 127.6 x 14.4 mm, Bottom Surface Area: 0.33cm2, Total Volume: 400µl, Working Volume: 350µl.