Learn More
Abcam Immunoprecipitation kit, 25 Tests
Catalog No. 502205085
Encompass_Preferred
Abcam's Immunoprecipitation Kit (ab206996) can be used to perform immunoprecipitation (IP) and Co-IP for functional studies of immunoprecipitated proteins/complexes and SDS-PAGE or western blot analysis of immunoprecipitated proteins and complexes.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-220-5085 Supplier Abcam Supplier No. AB20699625TEST
Abcam's Immunoprecipitation Kit (ab206996) can be used to perform immunoprecipitation (IP) and Co-IP for functional studies of immunoprecipitated proteins/complexes and SDS-PAGE or western blot analysis of immunoprecipitated proteins and complexes.
The product is subject to the following: Abcam Restricted Use Statement