Learn More
Apexbio Technology LLC Immunoprecipitation Kit (Protein A+G Agarose Gel) 20T
SDP
Catalog No. 5000097271
Encompass_Preferred
Immunoprecipitation Kit Protein AplusG Agarose Gel is a protein capture reagent designed to facilitate isolation of target proteins or protein-protein complexes from biological samples It utilizes Protein A G Agarose Gel which combines the binding profiles of Protein A and Protein G to interact specifically with the Fc regions of various antibodies This kit relies on the antibody-mediated capture of target proteins via Fc binding enabling selective enrichment and purification of individual proteins or their interaction partners from complex samples Based on these properties Immunoprecipitation Kit Protein AplusG Agarose Gel supports research applications such as analysis of protein interactions and enrichment of functional protein complexes for subsequent Western blot mass spectrometry or related downstream analyses
Catalog No. 50-000-97271 Supplier Apexbio Technology LLC Supplier No. K462320
Immunoprecipitation Kit Protein AplusG Agarose Gel is a protein capture reagent designed to facilitate isolation of target proteins or protein-protein complexes from biological samples It utilizes Protein A G Agarose Gel which combines the binding profiles of Protein A and Protein G to interact specifically with the Fc regions of various antibodies This kit relies on the antibody-mediated capture of target proteins via Fc binding enabling selective enrichment and purification of individual proteins or their interaction partners from complex samples Based on these properties Immunoprecipitation Kit Protein AplusG Agarose Gel supports research applications such as analysis of protein interactions and enrichment of functional protein complexes for subsequent Western blot mass spectrometry or related downstream analyses