Learn More
Apexbio Technology LLC Immunoprecipitation Kit (Protein G Agarose Gel) 20T
SDP
Catalog No. 5000097060
Encompass_Preferred
Immunoprecipitation Kit Protein G Agarose Gel is a reagent system based on Protein G agarose beads It is designed to enable selective isolation and analysis of proteins or protein complexes from biological samples using antibody-based affinity capture Immunoprecipitation Kit Protein G Agarose Gel functions by utilizing Protein G which specifically binds to the Fc region of various mammalian IgG subclasses thereby facilitating targeted immunoprecipitation (IP) and co-immunoprecipitation (Co-IP) of protein(s) of interest Based on these features Immunoprecipitation Kit Protein G Agarose Gel holds research potential in protein purification investigation of protein-protein interactions and downstream analyses such as Western blotting mass spectrometry and ELISA
Catalog No. 50-000-97060 Supplier Apexbio Technology LLC Supplier No. K462220
Immunoprecipitation Kit Protein G Agarose Gel is a reagent system based on Protein G agarose beads It is designed to enable selective isolation and analysis of proteins or protein complexes from biological samples using antibody-based affinity capture Immunoprecipitation Kit Protein G Agarose Gel functions by utilizing Protein G which specifically binds to the Fc region of various mammalian IgG subclasses thereby facilitating targeted immunoprecipitation (IP) and co-immunoprecipitation (Co-IP) of protein(s) of interest Based on these features Immunoprecipitation Kit Protein G Agarose Gel holds research potential in protein purification investigation of protein-protein interactions and downstream analyses such as Western blotting mass spectrometry and ELISA