Learn More
Bioworld Immunoprecipitation (IP) Buffer, 50 mL
Catalog No. 50198441
Encompass_Preferred
Immunoprecipitation bufferContains:NaCl, Tris-HCl pH-7.4, DTT, NP40, glycerol plus protease inhibitors.
Catalog No. 50-198-441 Supplier Bioworld Supplier No. 208310041
Immunoprecipitation bufferContains:NaCl, Tris-HCl pH-7.4, DTT, NP40, glycerol plus protease inhibitors.