Learn More
Bioworld Immunoprecipitation (IP) Buffer, 100 mL
Catalog No. 50198442
Encompass_Preferred
Immunoprecipitation bufferContains:NaCl, Tris-HCl pH-7.4, DTT, NP40, glycerol plus protease inhibitors.
Catalog No. 50-198-442 Supplier Bioworld Supplier No. 208310042
Immunoprecipitation bufferContains:NaCl, Tris-HCl pH-7.4, DTT, NP40, glycerol plus protease inhibitors.