Learn More
Beckman Coulter TQ-Prep, Sample Preparation Station
Catalog No. NC1908934
Encompass_Preferred
The Beckman Coulter TQ-Prep automates flow cytometry sample prep, standardizing pipetting, vortexing, and centrifugation. It features a compact design, intuitive interface, barcode scanning, and 21 CFR Part 11 compliance. Compatible with Navios and CytoFLEX systems, it enhances sample quality, reproducibility, and throughput.
Catalog No. NC1908934 Supplier Beckman Coulter Supplier No. 7546999
The Beckman Coulter TQ-Prep automates flow cytometry sample prep, standardizing pipetting, vortexing, and centrifugation. It features a compact design, intuitive interface, barcode scanning, and 21 CFR Part 11 compliance. Compatible with Navios and CytoFLEX systems, it enhances sample quality, reproducibility, and throughput.