Learn More
Electron Microscopy Sciences Immunosaver 100 ML
SDP
Catalog No. 5031427
Encompass_Preferred
Immunosaver allows for Immunostaining with quick and easy activation cell membranes and the nucleus. Immunosaver provides efficient antigen retrieval for successful immunostaining of a wide variety of antigens under optimized conditions. Protocols for both Light and Electron Microscopy may be found with this reagent.
Catalog No. 50-314-27 Supplier Electron Microscopy Sciences Supplier No. 64142
Immunosaver allows for Immunostaining with quick and easy activation cell membranes and the nucleus. Immunosaver provides efficient antigen retrieval for successful immunostaining of a wide variety of antigens under optimized conditions. Protocols for both Light and Electron Microscopy may be found with this reagent.