Learn More
Enzo Life Sciences Plate (IMMUNOSET pack) (96 wells)
SDP
Catalog No. 50658091
Encompass_Preferred
This IMMUNOSET plate rack is a convenient set of 5 clear 96 well high-binding polystyrene plates and 5 plate sealers. These are the same high quality plates used to validate all of our IMMUNOSET products. Use of these plates will provide optimal performance. The use of different types of plates will produce different results and may affect the sensitivity of the assay due to the type of plastic used. Shipping: Ambient. Long Term Storage: Ambient
Catalog No. 50-658-091 Supplier Enzo Life Sciences Supplier No. ADI801930
This IMMUNOSET plate rack is a convenient set of 5 clear 96 well high-binding polystyrene plates and 5 plate sealers. These are the same high quality plates used to validate all of our IMMUNOSET products. Use of these plates will provide optimal performance. The use of different types of plates will produce different results and may affect the sensitivity of the assay due to the type of plastic used. Shipping: Ambient. Long Term Storage: Ambient