Learn More
Ted Pella Inc Moisture Chamber, clear
SDP
Supplier:  Ted Pella Inc 21049
Encompass
This is a low-cost, high precision moisture chamber for ten slides. Every moisture chamber is divided into ten individual compartments with approximately 12.7mm (1/2") empty space between the compartments. When the chamber lid is closed, eight barrier dividers are placed into the empty spaces between the microscope slide compartments completely isolating all compartments. This is a very desirable feature when doing immunostaining. The microscope slides are placed on four pedestal posts and four corner posts each 11.5mm (0.460") high, thus raising the surface of the slides approximately 12.7mm (1/2") off the floor to keep the slides away from the water below and making the slides easily retrievable (either by hand or by forcep). The chamber is fabricated out of heavy-duty polystyrene with an air-tight design to keep moisture in. The chamber is designed to be stackable in order to save counter and/or refrigerator space.
Catalog No. NC9425474