Learn More
Research Products International Corp Incubation Tray for Immunostaining, 10 7/8 x 11 7/8 x 2 3/16 Inches
SDP
Catalog No. 50550290
Encompass
Specially designed tray for microwave enhanced immunostaining procedures. Built-in dividers provide channels that distribute water evenly under the slides. The water absorbs microwave energy and prevents overheating of samples. The technique allows a standard three step staining procedure to be performed in as little as fifteen minutes.
Optional lids reduce evaporation and protect samples.
Catalog No. 50-550-290 Supplier Research Products International Corp Supplier No. 248270
Specially designed tray for microwave enhanced immunostaining procedures. Built-in dividers provide channels that distribute water evenly under the slides. The water absorbs microwave energy and prevents overheating of samples. The technique allows a standard three step staining procedure to be performed in as little as fifteen minutes.
Optional lids reduce evaporation and protect samples.