Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Immunostics Hema-Screen™ SPECIFIC™ iFOBT Cassette Kit

Catalog No. 23038055 Shop All Immunostics Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-038-055 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-038-055 Supplier Immunostics Supplier No. HSGDCAS25
May include imposed supplier surcharges.
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

10 iFOBT cassettes and 10 buffer tubes or 25 iFOBT cassettes and 25 buffer tubes

Use for the rapid and qualitative determination of human hemoglobin in fecal samples by immunochemical methods. This kit for direct patient sampling uses a collection tube and iFOBT cassette.

  • Immunochemical fecal occult blood kit
  • No dietary restrictions or interfering substances
  • Collection system consists of a buffer tube with its integral spiral stick for specimen collection and easy-to-read iFOBT cassette
  • In-office CLIA waived procedure with test results in five minutes
  • Sensitivity level of 50μg Hb/g feces with 100% specificity for human hemoglobin
  • Recommended for use in screening for colorectal cancer or gastrointestinal bleeding, hospital monitoring for bleeding from in-patient population

Intended for professional and laboratory use only

Order Info

Up to 30-day specimen stability at room temperature

Specifications

Clia Complexity Waived
Type iFOBT Cassette Kit
For Use With (Application) Immunochemical fecal occult Blood test
Includes 25 Cassettes, 25 Buffer tubes, 1 product insert
Quantity Each
Test Time 5 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.