Learn More
Immunostics Hema-Screen SPECIFIC™ iFOBT Controls
Catalog No. 23038069 Shop All Immunostics Products
Click to view available options
Quantity:
Each
Catalog No. Quantity
23-038-069 Each
1 options
Catalog No. 23-038-069 Supplier Immunostics Supplier No. HSGDCON
Includes
  • 1 Positive Control
  • 1 Negative Control
  • Patient Instructions
External controls utilizing human hemoglobin to aid end user in verifying functionality of the immunoassay
Specifications
Type Hema Screen™ SPECIFIC™ Hemoglobin Control
For Use With (Application) Independently verify the functionality and performance of the test
Includes 1 Positive control, 1 negative control, 1 package insert
Quantity Each