Learn More
Agdia Inc ImmunoStrip for Tomato spotted wilt virus (TSWV), 25 strips & buffer filled bags
SDP
Catalog No. NC1644182
Encompass
ImmunoStrip tests require no equipment or expertise to run. Results are obtained in as little as a few minutes making them perfect for use in the field or greenhouse. ImmunoStrips for the detection of Tomato spotted wilt virus (TSWV) have been found to exhibit cross-reactivity with some isolates of Groundnut ringspot virus (GRSV) and Tomato chlorotic spot virus (TCSV). This test has also been reported to cross-react with Alstroemeria necrotic streak virus.
Catalog No. NC1644182 Supplier Agdia Inc Supplier No. ISK 39300/0025
ImmunoStrip tests require no equipment or expertise to run. Results are obtained in as little as a few minutes making them perfect for use in the field or greenhouse. ImmunoStrips for the detection of Tomato spotted wilt virus (TSWV) have been found to exhibit cross-reactivity with some isolates of Groundnut ringspot virus (GRSV) and Tomato chlorotic spot virus (TCSV). This test has also been reported to cross-react with Alstroemeria necrotic streak virus.