Learn More
Creative Biomart Alefacept, Recombinant human LFA3 protein, Fc tagged
Supplier:  Creative Biomart THP0152
Encompass_Preferred
Immunosuppressive dimeric fusion protein that consists of the extracellular CD2-binding portion of the human leukocyte function antigen-3 (LFA-3) linked to the Fc (hinge, CH2 and CH3 domains) portion of human IgG1. Produced by CHO cells, mW is 91.4 kD.
Catalog No. 50-208-4203