Learn More
Cell Signaling Technology Immunosuppressive Tumor Microenvironment IHC Antibody Sampler Kit, 1 Kit
SDP
Catalog No. 503131981
Encompass
The Immunosuppressive Tumor Microenvironment IHC Antibody Sampler Kit provides an economical means of detecting CD8+ T cells and various markers of immunosuppressive cell types and mechanisms in formalin-fixed paraffin-embedded human tumor tissue samples.
  • Includes antibodies for detecting CD8A, PD-L1, FoxP3, CD68, CD206/MRC1, and LRRC15
  • Features anti-rabbit IgG, HRP-linked secondary antibody for detection
  • Optimized for immunohistochemistry (paraffin) applications
  • Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and less than 0.02% sodium azide
Catalog No. 50-313-1981 Supplier Cell Signaling Technology Supplier No. 67524T
The Immunosuppressive Tumor Microenvironment IHC Antibody Sampler Kit provides an economical means of detecting CD8+ T cells and various markers of immunosuppressive cell types and mechanisms in formalin-fixed paraffin-embedded human tumor tissue samples.
  • Includes antibodies for detecting CD8A, PD-L1, FoxP3, CD68, CD206/MRC1, and LRRC15
  • Features anti-rabbit IgG, HRP-linked secondary antibody for detection
  • Optimized for immunohistochemistry (paraffin) applications
  • Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and less than 0.02% sodium azide