Learn More
Alkali Scientific ASI™ Immunotube, 12.00 x 75.00, 5mL volume, PP, Unibinding, 500/Cs
SDP
Catalog No. 503027643
Encompass_Preferred
ASI™ provides immunotube for diagnostics and immunological purposes. Immunotubes are often used for determining hormone levels, for example TSH (thyroid stimulating hormone). With a length of 75 mm and a diameter of 12 mmMaterials- PS : Commonly used and transparent. Withstands weak acids and bases.- PP : With high chemical resistance, it withstands acids and bases at room temperature and is harness.▫ Maxibinding : Modified polystyrene surface for higher binding capacity for proteins and other                          molecules with both hydrophilic and hydrophobic regions.                          (Recommended for most ELISA experiments)▫ Unibinding    : Suitable for adsorption of hydrophobic molecules. (Antigen ELISA) ▪ 2 type surface▪ For immunoassay (ELISA, RIA)▪ High optical quality
Catalog No. 50-302-7643 Supplier Alkali Scientific Supplier No. ITPPU05
ASI™ provides immunotube for diagnostics and immunological purposes. Immunotubes are often used for determining hormone levels, for example TSH (thyroid stimulating hormone). With a length of 75 mm and a diameter of 12 mmMaterials- PS : Commonly used and transparent. Withstands weak acids and bases.- PP : With high chemical resistance, it withstands acids and bases at room temperature and is harness.▫ Maxibinding : Modified polystyrene surface for higher binding capacity for proteins and other                          molecules with both hydrophilic and hydrophobic regions.                          (Recommended for most ELISA experiments)▫ Unibinding    : Suitable for adsorption of hydrophobic molecules. (Antigen ELISA) ▪ 2 type surface▪ For immunoassay (ELISA, RIA)▪ High optical quality