Learn More
Immy sōna Coccidioides Antibody Lateral Flow Assay, Qualitative detection, IgM and IgG, Serum & CSF, 50 tests, 30-60 min result, 2-25 °C storage
SDP
Catalog No. NC2266635
Encompass_Preferred
sōna Coccidioides Antibody Lateral Flow Assay, Qualitative detection, IgM and IgG, Serum & CSF, 50 tests, 30-60 min result, 2-25 °C storage
This item is not returnable. View return policy
Catalog No. NC2266635 Supplier Immy Supplier No. CTA2003
sōna Coccidioides Antibody Lateral Flow Assay, Qualitative detection, IgM and IgG, Serum & CSF, 50 tests, 30-60 min result, 2-25 °C storage