Learn More
TARGETMOL CHEMICALS INC IMP-1710 5MG
SDP
Catalog No. 502580132
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. IMP-1710 is a selective UCH-L1 inhibitor with an IC50 value of 38 nM in a fluorescence polarization assay. purity: 99%
Catalog No. 50-258-0132 Supplier TARGETMOL CHEMICALS INC Supplier No. T375915MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. IMP-1710 is a selective UCH-L1 inhibitor with an IC50 value of 38 nM in a fluorescence polarization assay. purity: 99%