Learn More
Medchemexpress LLC IMP-761 1mg | 1mg
SDP
Catalog No. 5000323752
Encompass_Preferred
IMP-761 is a human IgG4 monoclonal antibody (mAb) targeting CD223/LAG3 IMP-761 binds to D1 of human LAG32 Recommend Isotype Controls Human IgG4 kappa Isotype Control (HY-P99003)[1]
Catalog No. 50-003-23752 Supplier Medchemexpress LLC Supplier No. HYP9913011MG
IMP-761 is a human IgG4 monoclonal antibody (mAb) targeting CD223/LAG3 IMP-761 binds to D1 of human LAG32 Recommend Isotype Controls Human IgG4 kappa Isotype Control (HY-P99003)[1]