Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One IMP CT™ 96-well Protein Crystallization Plate
GREENER_CHOICE

Catalog No. 07000706 Shop All Greiner Bio-One Products
Change view
Click to view available options
Well Shape:
Conical Bottom (V-well)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Well Shape
07-000-706 Conical Bottom (V-well)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-000-706 Supplier Greiner Bio-One Supplier No. 673101
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use in microbatch crystallization. Greiner Bio-One IMP@CT™ 96-well Protein Crystallization Plate features conical wells that avoid spreading of droplets.

  • Low Profile
  • Polystyrene
  • Conical wells
  • Flat Bottom
TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Array 8 x 12
Color Clear
Product Type Crystallization Microplate
Material Polystyrene
No. of Wells 96
Sterility Non-sterile
Well Shape Conical Bottom (V-well)
For Use With (Application) Microbatch crystallization
Profile Low
Quantity 80/Cs.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.