Learn More
American Radiolabeled Chemicals Inc IMP DIAM 8-3H 1LC
SDP
Catalog No. 50901125
Encompass
Inosine 5'-8-3H monophosphate diammonium salt 1mCi in ethanol:water (1:1), 1 mCi/ml, S.A. 5-10 Ci/mmol, M.W. 348.2, Shipped in Dry Ice
Catalog No. 50-901-125 Supplier American Radiolabeled Chemicals Inc Supplier No. ART 17961MC
Add to Cart
Add to Cart
Request a Quote
Inosine 5'-8-3H monophosphate diammonium salt 1mCi in ethanol:water (1:1), 1 mCi/ml, S.A. 5-10 Ci/mmol, M.W. 348.2, Shipped in Dry Ice