Learn More
Aviva Systems Biology Corp IMP4 Antibody - C-terminal region (ARP65172_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65172_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IMP4. It was validated on Western Blot. . Alternate names: BXDC4, MGC19606ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-288-448