Learn More
Aviva Systems Biology Corp IMP5 Antibody - middle region (ARP53336_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53336_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IMP5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human IMP5. Alternate names: SPPL2c, IMP5ApplicationWB. Species reactivity: Human, Yeast
Catalog No. 50-014-61001