Learn More
Fritsch Milling And Sizing Inc Impact Bar PULVERISETTE 14 Premium Line
SDP
Catalog No. 501533976
Encompass
The FRITSCH impact bar for PULVERISETTE 14 premium line is ideal for very gentle and fast grinding of especially heat-sensitive materials such as powder coatings or plastics as well as for the smooth pre-crushing and fine comminution of hard-brittle to soft, fatty or samples with residual moisture. The corresponding impact rotor and a special sieve ring for the impact bar must be ordered separately.
Catalog No. 50-153-3976 Supplier Fritsch Milling And Sizing Inc Supplier No. 14.4470.00
The FRITSCH impact bar for PULVERISETTE 14 premium line is ideal for very gentle and fast grinding of especially heat-sensitive materials such as powder coatings or plastics as well as for the smooth pre-crushing and fine comminution of hard-brittle to soft, fatty or samples with residual moisture. The corresponding impact rotor and a special sieve ring for the impact bar must be ordered separately.